Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-O-Hexadecyl-2-N-methylcarbamyl-sn-glycero-3-phosphocholine, 98%


Quantity 5mg

Manufacturer: alfa aesar™  J67170LB0

 View more versions of this product

Catalog No. AAJ67170LB0

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 538.71
Assay 98%
Chemical Name or Material 1-O-Hexadecyl-2-N-methylcarbamyl-sn-glycero-3-phosphocholine
CAS 91575-58-5
Synonym Carbamyl-PAF
Physical Form Solid
Storage Note 1 -30° to -10°C
Color White
Quantity 5mg
MDL Number MFCD00065891

Specifications

Chemical Name or Material 1-O-Hexadecyl-2-N-methylcarbamyl-sn-glycero-3-phosphocholine
CAS 91575-58-5
Formula Weight 538.71
MDL Number MFCD00065891
Synonym Carbamyl-PAF
Accessories

-30° to -10°C

SDS & Safety Documents