Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-O-Hexadecyl-2-N-methylcarbamyl-sn-glycero-3-phosphocholine, 98%

Chemical Identifiers

Chemical Name or Material 1-O-Hexadecyl-2-N-methylcarbamyl-sn-glycero-3-phosphocholine
CAS 91575-58-5
Formula Weight 538.71
MDL Number MFCD00065891
Synonym Carbamyl-PAF
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ67170LB0

  SDS
alfa aesar™
J67170LB0
5mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAJ67170MB

  SDS
alfa aesar™
J67170MB
25mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 538.71
Chemical Name or Material 1-O-Hexadecyl-2-N-methylcarbamyl-sn-glycero-3-phosphocholine
Physical Form Solid
Assay 98%
CAS 91575-58-5
Synonym Carbamyl-PAF
Storage Note 1 -30° to -10°C
MDL Number MFCD00065891
Color White
Quantity 5mg
Safety and Handling

-30° to -10°C