missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
1-Nonyne 98.0+%, TCI America™
$74.65 - $74.65
Chemical Identifiers
CAS | 3452-09-3 |
---|---|
Molecular Formula | C9H16 |
Molecular Weight (g/mol) | 124.23 |
MDL Number | MFCD00009567 |
InChI Key | OSSQSXOTMIGBCF-UHFFFAOYSA-N |
Synonym | 1-nonyne, nonyne, unii-760v9gp5a1, 3-acetylnaphthalene, pubchem13060, acmc-1aglr, 1-nonyne 6ci, 7ci, 8ci, 9ci |
PubChem CID | 18937 |
IUPAC Name | non-1-yne |
SMILES | CCCCCCCC#C |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
N03015ML
|
TCI America
N03015ML |
5 mL |
Each for $74.65
|
|
|||||
Chemical Identifiers
3452-09-3 | |
124.23 | |
OSSQSXOTMIGBCF-UHFFFAOYSA-N | |
18937 | |
CCCCCCCC#C |
C9H16 | |
MFCD00009567 | |
1-nonyne, nonyne, unii-760v9gp5a1, 3-acetylnaphthalene, pubchem13060, acmc-1aglr, 1-nonyne 6ci, 7ci, 8ci, 9ci | |
non-1-yne |
Specifications
3452-09-3 | |
151°C | |
MFCD00009567 | |
3295 | |
OSSQSXOTMIGBCF-UHFFFAOYSA-N | |
non-1-yne | |
18937 | |
≥98.0% (GC) | |
1-Nonyne |
Colorless | |
C9H16 | |
5 mL | |
1-nonyne, nonyne, unii-760v9gp5a1, 3-acetylnaphthalene, pubchem13060, acmc-1aglr, 1-nonyne 6ci, 7ci, 8ci, 9ci | |
CCCCCCCC#C | |
124.23 | |
124.23 | |
Liquid |
Safety and Handling
TSCA : Yes