missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Nonyne 98%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC392750050

 View more versions of this product

Catalog No. AC392750050

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

≥97.5%
Liquid
Clear colorless to pale yellow
567104
CH3(CH2)6CCH
C9H16
97.5
Glass bottle
98%
124.23
5g
0.757g/mL
100.0
Authentic
1-Nonyne
MFCD00009567
1.4210 to 1.4230 (20°C, 589nm)

Chemical Identifiers

567104
100.0
C9H16
124.23
1-Nonyne
97.5
98%
CH3(CH2)6CCH
MFCD00009567
≥97.5%

Safety and Handling

SDS & Safety Documents

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Flammable liquid and vapor.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning