Please login to your online account to display your discounted pricing

1-Nonyne 98%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 1-Nonyne
CAS 567104
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C9H16
Linear Formula CH3(CH2)6CCH
Formula Weight 124.23
MDL Number MFCD00009567
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC392750050

  SDS
thermo fisher scientific
AC392750050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC392750250

  SDS
thermo fisher scientific
AC392750250
Glass bottle 25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥97.5%
Formula Weight 124.23
Physical Form Liquid
Quantity 5g
Color Clear colorless to pale yellow
Density 0.757g/mL
CAS 567104
CAS Max % 100.0
Linear Formula CH3(CH2)6CCH
Infrared Spectrum Authentic
Molecular Formula C9H16
Chemical Name or Material 1-Nonyne
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Flammable liquid and vapor.
CAS Min % 97.5
MDL Number MFCD00009567
Packaging Glass bottle
Refractive Index 1.4210 to 1.4230 (20°C, 589nm)
Assay Percent Range 98%
Health Hazard 1 GHS Signal Word: Warning
Safety and Handling

GHS Signal Word: Warning

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Flammable liquid and vapor.