Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Nonyne, 98%

Chemical Identifiers

Chemical Name or Material 1-Nonyne
CAS 3452-09-3
Molecular Formula C9H16
Linear Formula CH3(CH2)6C≡CH
Formula Weight 124.23
MDL Number MFCD00009567
UN Number UN3295
Beilstein 1736573
Synonym n-Heptylacetylene
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2426306

 
alfa aesar™
B2426306
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 124.23
Beilstein 1736573
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H304-H226
May be fatal if swallowed and enters airways.
Flammable liquid and vapour.
Boiling Point 149° to 151°C
Chemical Name or Material 1-Nonyne
TSCA TSCA
Assay 98%
Molecular Formula C9H16
Linear Formula CH3(CH2)6C≡CH
MDL Number MFCD00009567
CAS 3452-09-3
UN Number UN3295
Synonym n-Heptylacetylene
Health Hazard 3 GHS P Statement
P301+P330+P331-P315
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Get immediate medical advice/attention.
DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: HYDROCARBONS, LIQUID, N.O.S.
Flash Point 33°C (91°F)
Odor Petrol-like
Melting Point −50°C
Quantity 5g

Applications
1-Nonyne has been used in enantioselective synthesis of cladospolide B, C and (ent)-cladospolide D.

Solubility
Insoluble in water.

Notes
Store away from oxidizing agents, heavy metal powders.

Safety and Handling

Danger

GHS P Statement
P301+P330+P331-P315
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Get immediate medical advice/attention.

GHS H Statement
H304-H226
May be fatal if swallowed and enters airways.
Flammable liquid and vapour.