Please login to your online account to display your discounted pricing

Alfa Aesar™ (1-Nonyl)triphenylphosphonium bromide, 98+%

$45.81 - $45.81

Chemical Identifiers

Chemical Name or Material (1-Nonyl)triphenylphosphonium bromide
CAS 60902-45-6
Molecular Formula CH3(CH2)8P(C6H5)3Br
Formula Weight 469.45
MDL Number MFCD00051875
Beilstein 4223018
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0234106

  SDS
alfa aesar™
L0234106
5g Each for $45.81
Description & Specifications

Specifications

Formula Weight 469.45
Chemical Name or Material (1-Nonyl)triphenylphosphonium bromide
CAS 60902-45-6
Molecular Formula CH3(CH2)8P(C6H5)3Br
Health Hazard 1 Warning
TSCA TSCA
Assay ≥98%
Melting Point 63° to 65°C
MDL Number MFCD00051875
Beilstein 4223018
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 5g
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.