missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Nonyl isothiocyanate, 97%

Manufacturer:  Alfa Aesar™ L1006603

 View more versions of this product

Catalog No. AAL1006603



Specifications

Specifications

1-Nonyl isothiocyanate
119°C to 120°C (30mmHg)
4430-43-7
MFCD00041141
1761894
GSAYWRFRZHSPQB-UHFFFAOYSA-N
1-isothiocyanatononane
123217
97%
Pungent
1g
C10H19NS
UN2922
nonyl isothiocyanate, n-nonyl isothiocyanate, 1-nonyl isothiocyanate, isothiocyanic acid, nonyl ester, nonylisothiocyanate, acmc-1aiko, nonane,1-isothiocyanato
CCCCCCCCCN=C=S
185.329
185.33

Chemical Identifiers

4430-43-7
185.329
GSAYWRFRZHSPQB-UHFFFAOYSA-N
123217
CCCCCCCCCN=C=S
C10H19NS
MFCD00041141
nonyl isothiocyanate, n-nonyl isothiocyanate, 1-nonyl isothiocyanate, isothiocyanic acid, nonyl ester, nonylisothiocyanate, acmc-1aiko, nonane,1-isothiocyanato
1-isothiocyanatononane
Safety and Handling

Safety and Handling

GHS H Statement
H331-H314-H318-H302-H312
Toxic if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUIDS, TOXIC, N.O.S.

Documents