Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Nonen-4-ol, 98%

$23.66 - $90.09

Chemical Identifiers

Chemical Name or Material 1-Nonen-4-ol
CAS 35192-73-5
Molecular Formula C9H18O
Formula Weight 142.24
MDL Number MFCD00021949
Synonym Allyl n-pentyl carbinol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2028403

  SDS
alfa aesar™
B2028403
1g Each for $23.66

AAB2028406

  SDS
alfa aesar™
B2028406
5g Each for $90.09
Description & Specifications

Specifications

Formula Weight 142.24
Molecular Formula C9H18O
CAS 35192-73-5
Synonym Allyl n-pentyl carbinol
MDL Number MFCD00021949
Assay 98%
Quantity 1g
Chemical Name or Material 1-Nonen-4-ol