Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Nonen-3-ol, 98%

$72.93 - $245.61

Chemical Identifiers

Chemical Name or Material 1-Nonen-3-ol
CAS 21964-44-3
Molecular Formula C9H18O
Formula Weight 142.24
MDL Number MFCD00021953
Beilstein 1745976
Synonym n-Hexyl vinyl carbinol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0688906

  SDS
alfa aesar™
L0688906
5g Each for $72.93

AAL0688914

  SDS
alfa aesar™
L0688914
25g Each for $245.61
Description & Specifications

Specifications

Formula Weight 142.24
MDL Number MFCD00021953
Assay 98%
Chemical Name or Material 1-Nonen-3-ol
Health Hazard 2 GHS H Statement
H227
Combustible liquid.
Flash Point 84°C (183°F)
CAS 21964-44-3
Molecular Formula C9H18O
Beilstein 1745976
Synonym n-Hexyl vinyl carbinol
Health Hazard 1 Warning
Quantity 5g
TSCA TSCA
Boiling Point 192° to 194°C
Health Hazard 3 GHS P Statement
P210-P280-P370+P378a-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
In case of fire: Use for extinction: CO2, powder or water spray.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P210-P280-P370+P378a-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
In case of fire: Use for extinction: CO2, powder or water spray.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H227
Combustible liquid.