Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Nonanethiol, 98%

$63.11 - $207.80

Chemical Identifiers

Chemical Name or Material 1-Nonanethiol
CAS 1455-21-6
Molecular Formula CH3(CH2)8SH
Formula Weight 160.32
MDL Number MFCD00004913
Beilstein 1733631
Synonym n-Nonyl mercaptan
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1098814

 
alfa aesar™
L1098814
25g Each for $207.80

AAL1098806

 
alfa aesar™
L1098806
5g Each for $63.11
Description & Specifications

Specifications

Formula Weight 160.32
Chemical Name or Material 1-Nonanethiol
TSCA TSCA
Assay 98%
Molecular Formula CH3(CH2)8SH
MDL Number MFCD00004913
Beilstein 1733631
Synonym n-Nonyl mercaptan
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H227
Causes skin irritation.
Causes serious eye irritation.
Combustible liquid.
Health Hazard 3 GHS P Statement
P210-P280-P305+P351+P338-P362-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Take off contaminated clothing and wash before reuse.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Boiling Point 219° to 220°C
CAS 1455-21-6
Flash Point 79°C (174°F)
Quantity 25g
Safety and Handling

Warning

GHS P Statement
P210-P280-P305+P351+P338-P362-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Take off contaminated clothing and wash before reuse.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H227
Causes skin irritation.
Causes serious eye irritation.
Combustible liquid.