missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Nonanethiol, 98%

Manufacturer:  Alfa Aesar™ L1098806

 View more versions of this product

Catalog No. AAL1098806



Specifications

Specifications

1-Nonanethiol
219°C to 220°C
1455-21-6
MFCD00004913
1-nonanethiol, n-nonyl mercaptan, nonyl mercaptan, nonylthiol, unii-39lg2etv2j, 39lg2etv2j, nonanethiol, non-thiol, n-nonylmercaptan, 1-nonylthiol
CCCCCCCCCS
160.319
160.32
79°C (174°F)
5g
C9H20S
1733631
ZVEZMVFBMOOHAT-UHFFFAOYSA-N
nonane-1-thiol
15077
98%

Chemical Identifiers

1455-21-6
160.319
ZVEZMVFBMOOHAT-UHFFFAOYSA-N
15077
CCCCCCCCCS
C9H20S
MFCD00004913
1-nonanethiol, n-nonyl mercaptan, nonyl mercaptan, nonylthiol, unii-39lg2etv2j, 39lg2etv2j, nonanethiol, non-thiol, n-nonylmercaptan, 1-nonylthiol
nonane-1-thiol
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H227
Causes skin irritation.
Causes serious eye irritation.
Combustible liquid.

GHS P Statement
P210-P280-P305+P351+P338-P362-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Take off contaminated clothing and wash before reuse.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

TSCA : TSCA

Documents