Please login to your online account to display your discounted pricing

1-Nonanesulfonic Acid Sodium Salt, Anhydrous, +99%, ACROS Organics™


Quantity 25g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC396700250

 View more versions of this product

Catalog No. AC396700250

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥99.0% (alkal. after ion exchange) on dry substance
Physical Form Crystalline Powder
Formula Weight 230.3
Quantity 25g
Color White
Purity Grade Ion pair chromatography
CAS Min % 99.0
CAS Max % 100.0
Absorbance 0.02 max. (C=0.2 M in water) at 230nm
Suitability (HPLC, gradient) Passes
Infrared Spectrum Authentic
Linear Formula S(O3Na)(CH2)8CH3
Additional Information Membrane filtration test: Passes Test on 0.2M sol
Loss on Drying 2% max. (150°C)
Molecular Formula C9H19NaO3S
Packaging Glass bottle
MDL Number MFCD00025010
Chemical Name or Material 1-Nonanesulfonic acid, sodium salt
Assay Percent Range 99+%
Name Note Anhydrous
CAS 35192-74-6

Specifications

Chemical Name or Material 1-Nonanesulfonic acid, sodium salt
Name Note Anhydrous
CAS 35192-74-6
CAS Min % 99.0
CAS Max % 100.0
Purity Grade Ion pair chromatography
Assay ≥99.0% (alkal. after ion exchange) on dry substance
Assay Percent Range 99+%
Molecular Formula C9H19NaO3S
Linear Formula S(O3Na)(CH2)8CH3
Formula Weight 230.3
MDL Number MFCD00025010