Please login to your online account to display your discounted pricing

1-Nonanesulfonic Acid Sodium Salt, Anhydrous, +99%, ACROS Organics™

$110.42 - $110.42

Chemical Identifiers

Chemical Name or Material 1-Nonanesulfonic acid, sodium salt
Name Note Anhydrous
CAS 35192-74-6
CAS Min % 99.0
CAS Max % 100.0
Purity Grade Ion pair chromatography
Assay ≥99.0% (alkal. after ion exchange) on dry substance
Assay Percent Range 99+%
Molecular Formula C9H19NaO3S
Linear Formula S(O3Na)(CH2)8CH3
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC396700050

  SDS
thermo fisher scientific
AC396700050
Glass bottle 5g Each for $110.42

AC396700250

  SDS
thermo fisher scientific
AC396700250
Glass bottle 25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Crystalline Powder
Assay ≥99.0% (alkal. after ion exchange) on dry substance
Formula Weight 230.3
Quantity 5g
Color White
Purity Grade Ion pair chromatography
CAS Min % 99.0
CAS Max % 100.0
Absorbance 0.02 max. (C=0.2 M in water) at 230nm
Loss on Drying 2% max. (150°C)
Name Note Anhydrous
Molecular Formula C9H19NaO3S
Packaging Glass bottle
Additional Information Membrane filtration test: Passes Test on 0.2M sol
Suitability (HPLC, gradient) Passes
Linear Formula S(O3Na)(CH2)8CH3
Infrared Spectrum Authentic
Chemical Name or Material 1-Nonanesulfonic acid, sodium salt
Assay Percent Range 99+%
MDL Number MFCD00025010
CAS 35192-74-6

For ion pair chromatography