Please login to your online account to display your discounted pricing

1-Nonanesulfonic acid sodium salt, 97%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 1-Nonanesulfonic acid sodium salt
CAS 35192-74-6
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C9H19NaO3S
Linear Formula S(O3Na)(CH2)8CH3
Formula Weight 230.3
MDL Number MFCD00025010
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC338420010

  SDS
thermo fisher scientific
338420010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC338420050

  SDS
thermo fisher scientific
338420050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥96.0%
Physical Form Fine Powder
Formula Weight 230.3
Quantity 1g
Color White
CAS Max % 100.0
CAS Min % 96.0
Linear Formula S(O3Na)(CH2)8CH3
Infrared Spectrum Authentic
Assay Percent Range 97%
CAS 35192-74-6
Chemical Name or Material 1-Nonanesulfonic acid sodium salt
Molecular Formula C9H19NaO3S
MDL Number MFCD00025010
Packaging Glass bottle