missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Nitropyrazole, TCI America™

$223.06 - $666.13

Chemical Identifiers

CAS 7119-95-1
Molecular Formula C3H3N3O2
Molecular Weight (g/mol) 113.076
MDL Number MFCD00015894
InChI Key TYNVOQYGXDUHRX-UHFFFAOYSA-N
PubChem CID 146002
IUPAC Name 1-nitropyrazole
SMILES C1=CN(N=C1)[N+](=O)[O-]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
N09655G View Documents TCI America
N09655G
5g Each for $223.06
 
N096525G View Documents TCI America
N096525G
25g Each for $666.13
 
Specifications

Chemical Identifiers

7119-95-1
113.076
TYNVOQYGXDUHRX-UHFFFAOYSA-N
1-nitropyrazole
C3H3N3O2
MFCD00015894
146002
C1=CN(N=C1)[N+](=O)[O-]

Specifications

White-Yellow
5g
C3H3N3O2
TYNVOQYGXDUHRX-UHFFFAOYSA-N
1-nitropyrazole
146002
Crystal-Powder at 20°C
1-Nitropyrazole
94°C
7119-95-1
MFCD00015894
C1=CN(N=C1)[N+](=O)[O-]
113.076
113.08
≥97.0% (GC)
Safety and Handling

Safety and Handling

TSCA : No