missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Nitropropane 98%, ACROS Organics™

$29.47 - $90.90

Chemical Identifiers

CAS 108-03-2
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Molecular Formula C3H7NO2
Linear Formula CH3CH2CH2NO2
Formula Weight 89.09
MDL Number MFCD00007407
Chemical Name or Material 1-Nitropropane, 98%
Assay ≥97.5%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC128900050 SDS Thermo Fisher Scientific
128900050
Glass bottle 5g Each for $29.47
AC128905000 SDS Thermo Fisher Scientific
128905000
Glass bottle 500g Each for $90.90
Specifications Description & Specifications

Chemical Identifiers

108-03-2
100.0
C3H7NO2
89.09
1-Nitropropane, 98%
97.5
98%
CH3CH2CH2NO2
MFCD00007407
≥97.5%

Specifications

0.2% max.
97.5
Authentic
1.4005 to 1.4025 (20°C, 589nm)
0.99g/mL
98%
CH3CH2CH2NO2
89.09
5g
01,745
1-Nitropropane, 98%
108-03-2
100.0
Glass bottle
0.1% max.
Liquid
C3H7NO2
Clear
MFCD00007407
01, 115
15, 6714
≥97.5%

Safety and Handling

GHS H Statement:
Harmful in contact with skin.
Harmful if inhaled.
Harmful if swallowed.
Flammable liquid and vapor.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 203-544-9

RTECSNumber : TZ5075000

TSCA : TSCA