missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Nitropropane, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A1197530

 View more versions of this product

Catalog No. AAA1197530



Description

Description

The solvent of 1-Nitropropane is mainly used coatings, inks and separation processes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
The solvent of 1-Nitropropane is mainly used coatings, inks and separation processes.

Solubility
Soluble in water. (1.4 g/L)

Notes
Store in cool place. Keep away from oxidizing agents.
Specifications

Specifications

1-Nitropropane
-108°C
131°C to 132°C
Unpleasant
1.402
MFCD00007407
UN2608
14,6627
Soluble in water. (1.4g/L)
CCC[N+](=O)[O-]
89.094
CHEBI:76261
98%
108-03-2
0.998
34°C (93°F)
C3H7NO2
CH3CH2CH2NO2
250 g
506236
propane, 1-nitro, nitropropane, propane, nitro, ccris 1329, dsstox_cid_980, dsstox_rid_75902, dsstox_gsid_20980, nitro-propane, n-nitropropane, 3-nitropropane
JSZOAYXJRCEYSX-UHFFFAOYSA-N
1-nitropropane
7903
89.09

Chemical Identifiers

108-03-2
89.094
JSZOAYXJRCEYSX-UHFFFAOYSA-N
7903
1-nitropropane
C3H7NO2
MFCD00007407
propane, 1-nitro, nitropropane, propane, nitro, ccris 1329, dsstox_cid_980, dsstox_rid_75902, dsstox_gsid_20980, nitro-propane, n-nitropropane, 3-nitropropane
CHEBI:76261
CCC[N+](=O)[O-]
Safety and Handling

Safety and Handling

GHS H Statement
H226-H302-H312-H332
Flammable liquid and vapor.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.

P210-P233-P235-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P311-P312-P330-P363-P370+P378q-P501c

H226-H302+H312-H331

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: NITROPROPANES

EINECSNumber : 203-544-9

RTECSNumber : TZ5075000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only