Please login to your online account to display your discounted pricing

1-Nitronaphthalene 99%, ACROS Organics™

$64.84 - $64.84

Chemical Identifiers

Chemical Name or Material 1-Nitronaphthalene
Name Note 99%
CAS 86-57-7
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C10H7NO2
Linear Formula C10H7NO2
Formula Weight 173.17
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC147040050

  SDS
thermo fisher scientific
147040050
Glass bottle 5g Each for $64.84
Description & Specifications

Specifications

Physical Form Crystalline Powder and Chunks
Assay ≥98.5%
Formula Weight 173.17
Color Yellow to brownish
Quantity 5g
EINECS Number 201-684-5
TSCA TSCA
RTECS Number QJ9720000
Density 1.2200g/mL
Melting Point 56° to 59°C
Beilstein 05, 553
Merck Index 15, 6701
CAS Max % 100.0
Chemical Name or Material 1-Nitronaphthalene
Packaging Glass bottle
Name Note 99%
Assay Percent Range 99%
Molecular Formula C10H7NO2
Linear Formula C10H7NO2
MDL Number MFCD00003913
CAS 86-57-7
CAS Min % 98.5
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 2 GHS H Statement:
Toxic if swallowed.
Fatal if inhaled.
Suspected of causing cancer.
Flammable solid.
Health Hazard 3 GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Immediately call a POISON CENTER or doctor/physician.
Use personal protective equipment as required.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep container tightly closed.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Infrared Spectrum Authentic
Safety and Handling

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Immediately call a POISON CENTER or doctor/physician.
Use personal protective equipment as required.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep container tightly closed.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Toxic if swallowed.
Fatal if inhaled.
Suspected of causing cancer.
Flammable solid.