Please login to your online account to display your discounted pricing

1-Nitronaphthalene 99%, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  147040050

 View more versions of this product

Catalog No. AC147040050

  • / Each
 

Description & Specifications

Chemical Identifiers

Color Yellow to brownish
Formula Weight 173.17
Assay ≥98.5%
Physical Form Crystalline Powder and Chunks
Quantity 5g
TSCA TSCA
EINECS Number 201-684-5
RTECS Number QJ9720000
Melting Point 56° to 59°C
Density 1.2200g/mL
Beilstein 05, 553
Merck Index 15, 6701
CAS Max % 100.0
Packaging Glass bottle
Health Hazard 3 GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Immediately call a POISON CENTER or doctor/physician.
Use personal protective equipment as required.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep container tightly closed.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS H Statement:
Toxic if swallowed.
Fatal if inhaled.
Suspected of causing cancer.
Flammable solid.
CAS Min % 98.5
Infrared Spectrum Authentic
Health Hazard 1 GHS Signal Word: Danger
Linear Formula C10H7NO2
Molecular Formula C10H7NO2
Assay Percent Range 99%
MDL Number MFCD00003913
Name Note 99%
CAS 86-57-7
Chemical Name or Material 1-Nitronaphthalene

Specifications

Chemical Name or Material 1-Nitronaphthalene
Name Note 99%
CAS 86-57-7
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C10H7NO2
Linear Formula C10H7NO2
Formula Weight 173.17
MDL Number MFCD00003913
Accessories

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Immediately call a POISON CENTER or doctor/physician.
Use personal protective equipment as required.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep container tightly closed.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Toxic if swallowed.
Fatal if inhaled.
Suspected of causing cancer.
Flammable solid.

SDS & Safety Documents