Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Nitro-4-(trifluoromethoxy)benzene, 98+%

$41.55 - $358.65

Chemical Identifiers

Chemical Name or Material 1-Nitro-4-(trifluoromethoxy)benzene
CAS 713-65-5
Molecular Formula C7H4F3NO3
Formula Weight 207.11
MDL Number MFCD00040302
Beilstein 1966388
Synonym 4-(Trifluoromethoxy)nitrobenzene
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1978303

 
alfa aesar™
L1978303
1g Each for $41.55

AAL1978306

 
alfa aesar™
L1978306
5g Each for $112.51

AAL1978314

 
alfa aesar™
L1978314
25g Each for $358.65
Description & Specifications

Specifications

Formula Weight 207.11
Assay ≥98%
MDL Number MFCD00040302
Synonym 4-(Trifluoromethoxy)nitrobenzene
Chemical Name or Material 1-Nitro-4-(trifluoromethoxy)benzene
Molecular Formula C7H4F3NO3
CAS 713-65-5
Beilstein 1966388
Health Hazard 1 Warning
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Boiling Point 87° to 89°C (15mmHg)
Melting Point 15°C
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Quantity 1g

Applications
1-Nitro-4-(trifluoromethoxy)benzene is an important raw material and intermediate used in organic synthesis, pharmaceuticals agrochemicals and dyestuff fields.

Solubility
Not miscible or difficult to mix in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.