missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Nitro-3,5-bis(trifluoromethyl)benzene, 97+%

$187.80 - $187.80

Chemical Identifiers

CAS 328-75-6
Molecular Formula C8H3F6NO2
Formula Weight 259.11
MDL Number MFCD00000384
Beilstein 1998251
Synonym 3,5-Bis(trifluoromethyl)nitrobenzene
Chemical Name or Material 1-Nitro-3,5-bis(trifluoromethyl)benzene
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1679714 Alfa Aesar™
A1679714
25g Each for $187.80
AAA1679706 Alfa Aesar™
A1679706
5g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

328-75-6
259.11
1998251
1-Nitro-3,5-bis(trifluoromethyl)benzene
C8H3F6NO2
MFCD00000384
3,5-Bis(trifluoromethyl)nitrobenzene

Specifications

77°C (170°F)
25g
C8H3F6NO2
MFCD00000384
3,5-Bis(trifluoromethyl)nitrobenzene
≥97%
75° to 76°C (11mmHg)
328-75-6
259.11
1998251
1-Nitro-3,5-bis(trifluoromethyl)benzene

Safety and Handling

GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.

GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning