missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Nitro-2,5-bis(trifluoromethyl)benzene, 98%

Manufacturer:  Alfa Aesar™ H6426006

 View more versions of this product

Catalog No. AAH6426006



Specifications

Specifications

1-Nitro-2,5-bis(trifluoromethyl)benzene
1.4270
90°C (15mmHg)
5g
C8H3F6NO2
2508751
BZKVUOHHNCMTLH-UHFFFAOYSA-N
2-nitro-1,4-bis(trifluoromethyl)benzene
2734048
Liquid
1.570g/mL
Light Sensitive
Yellow
320-88-7
MFCD00236674
2,5-bis trifluoromethyl nitrobenzene, 2-nitro-1,4-bis trifluoromethyl benzene, 1,4-bis trifluoromethyl-2-nitrobenzene, 1-nitro-2,5-bis trifluoromethyl benzene, benzene, 2-nitro-1,4-bis trifluoromethyl, 2,5-bistrifluoromethylnitrobenzene, pubchem19738, 2-nitro-1,4-bis-trifluoromethyl-benzene, acmc-1bn6y, ksc496c8f
C1=CC(=C(C=C1C(F)(F)F)[N+](=O)[O-])C(F)(F)F
259.107
259.11
98%

Chemical Identifiers

320-88-7
259.107
BZKVUOHHNCMTLH-UHFFFAOYSA-N
2734048
C1=CC(=C(C=C1C(F)(F)F)[N+](=O)[O-])C(F)(F)F
C8H3F6NO2
MFCD00236674
2,5-bis trifluoromethyl nitrobenzene, 2-nitro-1,4-bis trifluoromethyl benzene, 1,4-bis trifluoromethyl-2-nitrobenzene, 1-nitro-2,5-bis trifluoromethyl benzene, benzene, 2-nitro-1,4-bis trifluoromethyl, 2,5-bistrifluoromethylnitrobenzene, pubchem19738, 2-nitro-1,4-bis-trifluoromethyl-benzene, acmc-1bn6y, ksc496c8f
2-nitro-1,4-bis(trifluoromethyl)benzene
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.

GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

SDS Documents