Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Nitro-2,5-bis(trifluoromethyl)benzene, 98%

Chemical Identifiers

Chemical Name or Material 1-Nitro-2,5-bis(trifluoromethyl)benzene
CAS 320-88-7
Formula Weight 259.11
MDL Number MFCD00236674
Beilstein 2508751
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6426006

  SDS
alfa aesar™
H6426006
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6426014

  SDS
alfa aesar™
H6426014
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 259.11
Chemical Name or Material 1-Nitro-2,5-bis(trifluoromethyl)benzene
Refractive Index 1.4270
Physical Form Liquid
Assay 98%
MDL Number MFCD00236674
CAS 320-88-7
Beilstein 2508751
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Health Hazard 3 GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Density 1.570g/mL
Sensitivity Light Sensitive
Boiling Point 90°C (15mmHg)
Color Pale yellow to yellow
Quantity 5g
Safety and Handling

Warning

GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.