Please login to your online account to display your discounted pricing

Alfa Aesar™ (1-Naphthylmethyl)triphenylphosphonium chloride, 95%

$285.58 - $285.58

Chemical Identifiers

Chemical Name or Material (1-Naphthylmethyl)triphenylphosphonium chloride
CAS 23277-00-1
Molecular Formula C29H24ClP
Formula Weight 438.93g/mol
MDL Number MFCD00031658
Beilstein 3581515
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0066614

 
alfa aesar™
L00666-14
25g Each for $285.58
Description & Specifications

Specifications

Melting Point >300°C
Quantity 25g
Chemical Name or Material (1-Naphthylmethyl)triphenylphosphonium chloride
CAS 23277-00-1
Assay 95%
Molecular Formula C29H24ClP
Formula Weight 438.93g/mol
MDL Number MFCD00031658
Beilstein 3581515
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.