missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ (1-Naphthylmethyl)boronic acid pinacol ester, 96%

$240.57 - $718.62

Chemical Identifiers

CAS 475250-57-8
Molecular Formula C17H21BO2
Molecular Weight (g/mol) 268.163
MDL Number MFCD22419278
InChI Key VYMBZQMGIGIALJ-UHFFFAOYSA-N
Synonym 4,4,5,5-tetramethyl-2-naphthalen-1-ylmethyl-1,3,2-dioxaborolane, naphthalen-1-ylmethyl boronic acid pinacol ester, 2-1-naphthylmethyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 1-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl methyl naphthalene
PubChem CID 11254165
IUPAC Name 4,4,5,5-tetramethyl-2-(naphthalen-1-ylmethyl)-1,3,2-dioxaborolane
SMILES B1(OC(C(O1)(C)C)(C)C)CC2=CC=CC3=CC=CC=C32
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH62764MD View Documents Alfa Aesar™
H62764MD
250mg Each for $240.57
 
AAH6276403 View Documents Alfa Aesar™
H6276403
1g Each for $718.62
 
Description

Description

Applications
(1-Naphthylmethyl)boronic acid pinacol ester is employed as a intermediate for chemical research.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
Specifications

Chemical Identifiers

475250-57-8
268.163
VYMBZQMGIGIALJ-UHFFFAOYSA-N
11254165
B1(OC(C(O1)(C)C)(C)C)CC2=CC=CC3=CC=CC=C32
C17H21BO2
MFCD22419278
4,4,5,5-tetramethyl-2-naphthalen-1-ylmethyl-1,3,2-dioxaborolane, naphthalen-1-ylmethyl boronic acid pinacol ester, 2-1-naphthylmethyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 1-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl methyl naphthalene
4,4,5,5-tetramethyl-2-(naphthalen-1-ylmethyl)-1,3,2-dioxaborolane

Specifications

250mg
C17H21BO2
4,4,5,5-tetramethyl-2-naphthalen-1-ylmethyl-1,3,2-dioxaborolane, naphthalen-1-ylmethyl boronic acid pinacol ester, 2-1-naphthylmethyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 1-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl methyl naphthalene
B1(OC(C(O1)(C)C)(C)C)CC2=CC=CC3=CC=CC=C32
268.163
268.16
(1-Naphthylmethyl)boronic acid pinacol ester
475250-57-8
MFCD22419278
VYMBZQMGIGIALJ-UHFFFAOYSA-N
4,4,5,5-tetramethyl-2-(naphthalen-1-ylmethyl)-1,3,2-dioxaborolane
11254165
96%