Please login to your online account to display your discounted pricing

1-Naphthylmagnesium bromide, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 1-Naphthylmagnesium bromide
Name Note 0.25M solution in THF
CAS 703-55-9
CAS Min % 6.37
CAS Max % 6.63
CAS 109-99-9
CAS Min % 93.38
CAS Max % 93.63
Molecular Formula C10H7BrMg
Linear Formula C10H7MgBr
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC384121000

 
thermo fisher scientific
384121000
AcroSeal™ Glass Bottle in Tin Can 100mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Packaging AcroSeal™ Glass Bottle in Tin Can
Density 0.9080g/mL
Color Off-white to light brown
Quantity 100mL
Chemical Name or Material 1-Naphthylmagnesium bromide
Name Note 0.25M solution in THF
CAS 703-55-9
CAS Min % 6.37
CAS Max % 6.63
CAS 109-99-9
CAS Min % 93.38
CAS Max % 93.63
Physical Form Off-white to light brown powder
Molecular Formula C10H7BrMg
Linear Formula C10H7MgBr
Formula Weight 231.37g/mol
MDL Number MFCD00015768
Health Hazard 1 Danger
Health Hazard 2 GHS P Statement Causes severe skin burns and eye damage. May cause respiratory irritation. In contact with water releases flammable gases which may ignite spontaneously. Highly flammable liquid and vapor. May form exp
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Store in a dry place. Store in a closed container.
Keep container tightly closed.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.
Take precautionary measures against static discharge.
Store in a well-ventilated place. Keep cool.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Store in a dry place. Store in a closed container.
Keep container tightly closed.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.
Take precautionary measures against static discharge.
Store in a well-ventilated place. Keep cool.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS P Statement Causes severe skin burns and eye damage. May cause respiratory irritation. In contact with water releases flammable gases which may ignite spontaneously. Highly flammable liquid and vapor. May form exp