Please login to your online account to display your discounted pricing

1-Naphthylmagnesium bromide, ACROS Organics™


Quantity 100mL
Packaging AcroSeal™ Glass Bottle in Tin Can

Manufacturer: thermo fisher scientific  AC384121000

 View more versions of this product

Catalog No. AC384121000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 231.37
Physical Form Powder
Quantity 100mL
Color Off-white to light brown
Packaging AcroSeal™ Glass Bottle in Tin Can
Density 0.9080g/mL
Chemical Name or Material 1-Naphthylmagnesium bromide
Name Note 0.25M solution in THF
Molecular Formula C10H7BrMg
CAS 703-55-9
Health Hazard 1 Danger
CAS 109-99-9
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Store in a dry place. Store in a closed container.
Keep container tightly closed.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.
Take precautionary measures against static discharge.
Store in a well-ventilated place. Keep cool.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
CAS Max % 93.63
Health Hazard 2 GHS P Statement Causes severe skin burns and eye damage. May cause respiratory irritation. In contact with water releases flammable gases which may ignite spontaneously. Highly flammable liquid and vapor. May form exp
CAS Min % 93.38
CAS Min % 6.37
Linear Formula C10H7MgBr
MDL Number MFCD00015768
CAS Max % 6.63

Specifications

Chemical Name or Material 1-Naphthylmagnesium bromide
Name Note 0.25M solution in THF
CAS 703-55-9
CAS Min % 6.37
CAS Max % 6.63
CAS 109-99-9
CAS Min % 93.38
CAS Max % 93.63
Molecular Formula C10H7BrMg
Linear Formula C10H7MgBr
Formula Weight 231.37
MDL Number MFCD00015768
Accessories

Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Store in a dry place. Store in a closed container.
Keep container tightly closed.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.
Take precautionary measures against static discharge.
Store in a well-ventilated place. Keep cool.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS P Statement Causes severe skin burns and eye damage. May cause respiratory irritation. In contact with water releases flammable gases which may ignite spontaneously. Highly flammable liquid and vapor. May form exp