Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Naphthylmagnesium bromide, 0.5M in MeTHF

Chemical Identifiers

Chemical Name or Material 1-Naphthylmagnesium bromide
CAS 703-55-9
Molecular Formula C10H7BrMg
Formula Weight 231.37
MDL Number MFCD00015768
UN Number UN2924
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH51159AD

  SDS
alfa aesar™
H51159AD
50mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH51159AE

  SDS
alfa aesar™
H51159AE
100mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 231.37
Quantity 50mL
DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, CORROSIVE, N.O.S.
MDL Number MFCD00015768
Health Hazard 3 GHS P Statement
P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
CAS 703-55-9
Concentration 0.5M in MeTHF
Molecular Formula C10H7BrMg
Health Hazard 1 Danger
Chemical Name or Material 1-Naphthylmagnesium bromide
TSCA TSCA
UN Number UN2924
Health Hazard 2 GHS H Statement
H225-H314-H318
Highly flammable liquid and vapor.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Safety and Handling

Danger

GHS P Statement
P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H225-H314-H318
Highly flammable liquid and vapor.
Causes severe skin burns and eye damage.
Causes serious eye damage.