missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Naphthylhydrazine Hydrochloride, TCI America™

$79.45 - $268.89

Chemical Identifiers

CAS 2243-56-3
Molecular Formula C10H11ClN2
Molecular Weight (g/mol) 194.662
MDL Number MFCD00060157
InChI Key FYSSYOCJFZSKNW-UHFFFAOYSA-N
PubChem CID 519949
IUPAC Name naphthalen-1-ylhydrazine;hydrochloride
SMILES C1=CC=C2C(=C1)C=CC=C2NN.Cl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
N00641G View Documents TCI America
N00641G
1g Each for $79.45
 
N00645G View Documents TCI America
N00645G
5g Each for $268.89
 
Specifications

Chemical Identifiers

2243-56-3
194.662
FYSSYOCJFZSKNW-UHFFFAOYSA-N
naphthalen-1-ylhydrazine;hydrochloride
C10H11ClN2
MFCD00060157
519949
C1=CC=C2C(=C1)C=CC=C2NN.Cl

Specifications

Undesignated
2243-56-3
MFCD00060157
C1=CC=C2C(=C1)C=CC=C2NN.Cl
194.662
194.66
≥90.0% (HPLC)
1g
C10H11ClN2
FYSSYOCJFZSKNW-UHFFFAOYSA-N
naphthalen-1-ylhydrazine;hydrochloride
519949
Crystal-Powder at 20°C
1-Naphthylhydrazine Hydrochloride
Safety and Handling

Safety and Handling

TSCA : Yes