Please login to your online account to display your discounted pricing

1-Naphthylacetonitrile 97%, ACROS Organics™


Quantity 5g
Packaging Plastic bottle

Manufacturer: thermo fisher scientific  128230050

 View more versions of this product

Catalog No. AC128230050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥96.0%
Physical Form Liquid After Melting
Formula Weight 167.21
Quantity 5g
Color Clear yellow to brown
EINECS Number 205-078-1
TSCA TSCA
RTECS Number AM1240000
Beilstein 09, 667
CAS Min % 96.0
CAS Max % 100.0
Packaging Plastic bottle
Synonym 1-Naphthaleneacetonitrile
Health Hazard 1 GHS Signal Word: Warning
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Harmful if inhaled.
Harmful in contact with skin.
Chemical Name or Material 1-Naphthylacetonitrile
Refractive Index 1.6182 to 1.6202 (20°C, 589nm)
Molecular Formula C12H9N
Linear Formula C10H7CH2CN
MDL Number MFCD00004041
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Infrared Spectrum Authentic
Name Note 97%
Assay Percent Range 97%
CAS 132-75-2

Specifications

Chemical Name or Material 1-Naphthylacetonitrile
Name Note 97%
CAS 132-75-2
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C12H9N
Linear Formula C10H7CH2CN
Formula Weight 167.21
MDL Number MFCD00004041
Synonym 1-Naphthaleneacetonitrile
Accessories

GHS Signal Word: Warning

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.
Harmful if inhaled.
Harmful in contact with skin.

SDS & Safety Documents