Please login to your online account to display your discounted pricing

1-Naphthylacetonitrile 97%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 1-Naphthylacetonitrile
Name Note 97%
CAS 132-75-2
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C12H9N
Linear Formula C10H7CH2CN
Formula Weight 167.21
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC128230250

  SDS
thermo fisher scientific
128230250
Plastic bottle 25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC128230050

  SDS
thermo fisher scientific
128230050
Plastic bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥96.0%
Physical Form Liquid After Melting
Formula Weight 167.21
Quantity 25g
Color Clear yellow to brown
Beilstein 09, 667
EINECS Number 205-078-1
TSCA TSCA
RTECS Number AM1240000
CAS Min % 96.0
CAS Max % 100.0
Chemical Name or Material 1-Naphthylacetonitrile
Packaging Plastic bottle
Refractive Index 1.6182 to 1.6202 (20°C, 589nm)
Name Note 97%
Assay Percent Range 97%
Molecular Formula C12H9N
Linear Formula C10H7CH2CN
MDL Number MFCD00004041
CAS 132-75-2
Synonym 1-Naphthaleneacetonitrile
Health Hazard 1 GHS Signal Word: Warning
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Harmful if inhaled.
Harmful in contact with skin.
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Infrared Spectrum Authentic
Safety and Handling

GHS Signal Word: Warning

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.
Harmful if inhaled.
Harmful in contact with skin.