Please login to your online account to display your discounted pricing

1-Naphthylacetic acid, 95%, ACROS Organics™


Quantity 100g
Packaging Glass bottle

Manufacturer: fisher scientific acros  349641000

 View more versions of this product

Catalog No. AC349641000

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥94%
Physical Form Powder
Formula Weight 186.21
Quantity 100g
Color White to light yellow or beige
Beilstein 09, 666
Merck Index 15, 6456
Melting Point 126° to 133°C
CAS Min % 94.0
CAS Max % 100.0
Chemical Name or Material 1-Naphthylacetic acid
Assay Percent Range 95%
Molecular Formula C12H10O2
CAS 86-87-3
Synonym 1-Naphthaleneacetic acid
Health Hazard 1 Warning
Health Hazard 3 GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Packaging Glass bottle
MDL Number MFCD00004046
Infrared Spectrum Authentic
Health Hazard 2 GHS H Statement:
Harmful if swallowed.

Specifications

Chemical Name or Material 1-Naphthylacetic acid
CAS 86-87-3
CAS Min % 94.0
CAS Max % 100.0
Assay ≥94%
Assay Percent Range 95%
Molecular Formula C12H10O2
Formula Weight 186.21
MDL Number MFCD00004046
Accessories

Warning

GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.

SDS & Safety Documents