missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Naphthyl trifluoromethanesulfonate, 97%

Manufacturer:  Alfa Aesar™ H5509103

 View more versions of this product

Catalog No. AAH5509103



Description

Description

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
Specifications

Specifications

1-Naphthyl trifluoromethanesulfonate
113°C (235°F)
97°C to 98°C (0.3mmHg)
1g
C11H7F3O3S
UN3265
WQWUQDVFRYMMCY-UHFFFAOYSA-N
naphthalen-1-yl trifluoromethanesulfonate
378884
Liquid
1.405 g/mL
1.52
Red
99747-74-7
MFCD00192340
1-naphthalenyl triflate
C1=CC=C2C(=C1)C=CC=C2OS(=O)(=O)C(F)(F)F
276.229
276.23
97%

Chemical Identifiers

99747-74-7
276.229
WQWUQDVFRYMMCY-UHFFFAOYSA-N
378884
C1=CC=C2C(=C1)C=CC=C2OS(=O)(=O)C(F)(F)F
C11H7F3O3S
MFCD00192340
1-naphthalenyl triflate
naphthalen-1-yl trifluoromethanesulfonate
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314-H335

DOTInformation : Transport Hazard Class: 8; Packaging Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.

TSCA : No

Recommended Storage : Ambient temperatures

Documents