Please login to your online account to display your discounted pricing

1-Naphthyl PP1, Tocris

Src family kinase inhibitor; also inhibits c-Abl

$321.56 - $1,327.32

Chemical Identifiers

Chemical Name or Material 1-Naphthyl PP1
Assay Percent Range >99%
Molecular Formula C19H19N5
Formula Weight 317.39
Synonym 1-(1, 1-Dimethylethyl)-3-(1-naphthalenyl)-1H-pyrazolo[3, 4-d]pyrimidin-4-amine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

306310

 
r & d systems
306310
10mg Each for $321.56

306350

 
r & d systems
306350
50mg Each for $1,327.32
Description & Specifications

Specifications

Formula Weight 317.39
Quantity 10mg
Assay Percent Range >99%
Chemical Name or Material 1-Naphthyl PP1
CAS 221243-82-9
Molecular Formula C19H19N5
Synonym 1-(1, 1-Dimethylethyl)-3-(1-naphthalenyl)-1H-pyrazolo[3, 4-d]pyrimidin-4-amine
Recommended Storage Store at +4°C
Safety and Handling

Store at +4°C