missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Naphthyl phosphate, monosodium salt monohydrate, 98+%, ACROS Organics™

Manufacturer:  Fisher Scientific Acros 226890050

 View more versions of this product

Catalog No. AC226890050



Specifications

Specifications

(5% aq. soln.) clear, colorless
Authentic
White
5g
98+%
98.0
98+%
C10H7OP(O)(OH)ONa·H2O
1-naphthyl phosphate monosodium salt monohydrate, 1-naphthyl phosphate monosodium salt monohydrate hplc, 1-naphthyl phosphate sodium salt monohydrate, 1-naphthyl phosphate, monosodium salt monohydrate, alpha-naphthyl acid phosphate monosodium, c10h8po4.na.h2o, naphthalen-1-yloxy sodiooxy phosphinic acid hydrate, sodium 1-naphthyl phosphate monohydrate, sodium hydrate naphthalen-1-yl hydrogen phosphate, sodium naphthalen-1-yl hydrogenphosphate hydrate
OPNXTLGVPHOLCQ-UHFFFAOYSA-N
naphthalen-1-yl dihydrogen phosphate;sodium;hydrate
45055387
Fine Powder
1-Naphthyl phosphate, monosodium salt monohydrate
Glass bottle
189.0°C to 191.0°C
3.5 to 4.5 (5% aq. soln.)
81012-89-7
100.0
C10H8NaO4P·H2O
MFCD00150615
(5% in water) Clear colorless to light yellow
C1=CC=C2C(=C1)C=CC=C2OP(=O)(O)O.O.[Na]
264.15
264.15
98+%

Chemical Identifiers

81012-89-7
264.15
OPNXTLGVPHOLCQ-UHFFFAOYSA-N
45055387
C1=CC=C2C(=C1)C=CC=C2OP(=O)(O)O.O.[Na]
C10H8NaO4P·H2O
MFCD00150615
1-naphthyl phosphate monosodium salt monohydrate, 1-naphthyl phosphate monosodium salt monohydrate hplc, 1-naphthyl phosphate sodium salt monohydrate, 1-naphthyl phosphate, monosodium salt monohydrate, alpha-naphthyl acid phosphate monosodium, c10h8po4.na.h2o, naphthalen-1-yloxy sodiooxy phosphinic acid hydrate, sodium 1-naphthyl phosphate monohydrate, sodium hydrate naphthalen-1-yl hydrogen phosphate, sodium naphthalen-1-yl hydrogenphosphate hydrate
naphthalen-1-yl dihydrogen phosphate;sodium;hydrate
Safety and Handling

Safety and Handling

GHS H Statement
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get med

GHS Signal Word: Warning

TSCA : TSCA

SDS Documents