Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Naphthyl phosphate disodium salt hydrate, 99%

$185.34 - $718.73

Chemical Identifiers

Chemical Name or Material 1-Naphthyl phosphate disodium salt hydrate
CAS 207569-06-0
Molecular Formula C10H7Na2O4P·xH2O
Formula Weight 268.12 (Anhydrous)
MDL Number MFCD00150614
Beilstein 4895811
Synonym Disodium 1-naphthyl phosphate hydrate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2286614

  SDS
alfa aesar™
B2286614
25g Each for $718.73

AAB2286606

  SDS
alfa aesar™
B2286606
5g Each for $185.34
Description & Specifications

Specifications

Formula Weight 268.12 (Anhydrous)
Chemical Name or Material 1-Naphthyl phosphate disodium salt hydrate
Assay 99%
Molecular Formula C10H7Na2O4P·xH2O
Melting Point >300°C
MDL Number MFCD00150614
CAS 207569-06-0
Beilstein 4895811
Quantity 25g
Synonym Disodium 1-naphthyl phosphate hydrate
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Recommended Storage 2° to 8°C

Applications
1-Naphthyl phosphate disodium salt hydrate is used as an organic chemical synthesis intermediate.

Solubility
Soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents and heat.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

2° to 8°C