Please login to your online account to display your discounted pricing

1-Naphthyl phosphate, disodium salt hydrate, 98%, ACROS Organics™


Quantity 25g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC398260250

 View more versions of this product

Catalog No. AC398260250

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 268.11
Quantity 25g
CAS Max % 100.0
Chemical Name or Material 1-Naphthyl phosphate, disodium salt hydrate
MDL Number MFCD00150614
CAS 207569-06-0
CAS 2183-17-7
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Assay Percent Range 98%
Molecular Formula C10H7Na2O4P·xH2O
CAS Min % 97.5
Packaging Glass bottle
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

Specifications

Chemical Name or Material 1-Naphthyl phosphate, disodium salt hydrate
CAS 207569-06-0
CAS Min % 97.5
CAS Max % 100.0
CAS 2183-17-7
Assay Percent Range 98%
Molecular Formula C10H7Na2O4P·xH2O
Formula Weight 268.11
MDL Number MFCD00150614
Accessories

Warning

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

SDS & Safety Documents