missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Naphthyl phosphate, disodium salt hydrate, 98%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC398260010

 View more versions of this product

Catalog No. AC398260010

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

268.11
100.0
207569-06-0,2183-17-7
MFCD00150614
C10H7Na2O4P·xH2O
1g
1-Naphthyl phosphate, disodium salt hydrate
97.5
98%
Glass bottle

Chemical Identifiers

207569-06-0,2183-17-7
100.0
C10H7Na2O4P·xH2O
MFCD00150614
97.5
98%
268.11
1-Naphthyl phosphate, disodium salt hydrate

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning