Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Naphthyl isothiocyanate, 98%

Chemical Identifiers

Chemical Name or Material 1-Naphthyl isothiocyanate
CAS 551-06-4
Molecular Formula C11H7NS
Formula Weight 185.25
MDL Number MFCD00003882
UN Number UN2923
Beilstein 637868
Merck Index 14,6411
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1241809

  SDS
alfa aesar™
L1241809
10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 185.25
Chemical Name or Material 1-Naphthyl isothiocyanate
Assay 98%
Molecular Formula C11H7NS
MDL Number MFCD00003882
UN Number UN2923
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301-H311-H331-H314-H318
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Health Hazard 3 GHS P Statement
P260-P301+P310-P303+P361+P353-P305+P351+P338-P361-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLIDS, TOXIC, N.O.S.
TSCA TSCA
Melting Point 54° to 57°C
CAS 551-06-4
Beilstein 637868
Quantity 10g
Merck Index 14,6411
Odor Irritating
RTECS Number NX9100000

Applications
1-Naphthyl isothiocyanate is a tool for the study of liver damage which causes bile stasis and hyperbilirubinemia acutely and bile duct hyperplasia and biliary cirrhosis chronically, with changes in hepatocyte function.

Solubility
Hydrolyzes in water.

Notes
Moisture Sensitive. Incompatible with bases, alcohols and amines. Store under dry inert gas. Protect from humidity and water. Store away from moisture.

Safety and Handling

Danger

GHS P Statement
P260-P301+P310-P303+P361+P353-P305+P351+P338-P361-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H311-H331-H314-H318
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.