missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Naphthyl isocyanate, 99%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 415410050

 View more versions of this product

Catalog No. AC415410050



Description

Description

Specifications

Specifications

1-Naphthyl isocyanate
135°C
1.6324 to 1.6344
269°C to 270°C
5g
98.5
99%
15, 6492
Solubility in water: decomposes. Other solubilities: soluble in toluene,chloroform,ether,decomposes in alcohols
C1=CC=C2C(=C1)C=CC=C2N=C=O
169.18
169.18
1.1800g/mL
Glass bottle
1.18
4°C
86-84-0
100.0
C11H7NO
1-isocyanato-naphthalene, 1-isocyanatonapthalene, 1-naphthyl isocyanate, 1-naphthylisocyanate, alpha-naphthyl isocyanate, isocyanatonaphthalene, isocyanic acid 1-naphthyl ester, isocyanic acid, 1-naphthyl ester, naphthalene, 1-isocyanato, unii-5lh2p0691e
BDQNKCYCTYYMAA-UHFFFAOYSA-N
1-isocyanatonaphthalene
66589
99%

Chemical Identifiers

86-84-0
169.18
1-isocyanato-naphthalene, 1-isocyanatonapthalene, 1-naphthyl isocyanate, 1-naphthylisocyanate, alpha-naphthyl isocyanate, isocyanatonaphthalene, isocyanic acid 1-naphthyl ester, isocyanic acid, 1-naphthyl ester, naphthalene, 1-isocyanato, unii-5lh2p0691e
1-isocyanatonaphthalene
C11H7NO
BDQNKCYCTYYMAA-UHFFFAOYSA-N
66589
C1=CC=C2C(=C1)C=CC=C2N=C=O
Safety and Handling

Safety and Handling

GHS H Statement:
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Harmful in contact with skin.
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Harmful if swallowed.
Harmful if inhaled.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: If breathing is difficult, remove to fresh air and keep at rest in a position comfortable for breathing.
If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 201-703-7

SDS Documents