Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Naphthyl butyrate, 98+%

$48.07 - $461.07

Chemical Identifiers

Chemical Name or Material 1-Naphthyl butyrate
CAS 3121-70-8
Molecular Formula C14H14O2
Formula Weight 214.27
MDL Number MFCD00003923
Beilstein 2558646
Synonym Butyric acid 1-naphthyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2322103

  SDS
alfa aesar™
B2322103
1g Each for $48.07

AAB2322114

  SDS
alfa aesar™
B2322114
25g Each for $461.07
Description & Specifications

Specifications

Formula Weight 214.27
Molecular Formula C14H14O2
Boiling Point 125° to 130°C (1mmHg)
Quantity 1g
MDL Number MFCD00003923
TSCA TSCA
Chemical Name or Material 1-Naphthyl butyrate
Synonym Butyric acid 1-naphthyl ester
Flash Point 62°C (143°F)
Assay ≥98%
Health Hazard 2 GHS H Statement
H227
Combustible liquid.
Beilstein 2558646
Health Hazard 3 GHS P Statement
P210-P280-P370+P378a-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
In case of fire: Use for extinction: CO2, powder or water spray.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.
CAS 3121-70-8
Health Hazard 1 Warning
Safety and Handling

Warning

GHS P Statement
P210-P280-P370+P378a-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
In case of fire: Use for extinction: CO2, powder or water spray.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H227
Combustible liquid.