missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Naphthyl acetate, 99%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 227251000

 View more versions of this product

Catalog No. AC227251000



Specifications

Specifications

1-Naphthyl acetate
Glass bottle
43.0°C to 46.0°C
99+%
99.0
98.5% min. (HPLC)
CH3CO2C10H7
06, 608
VGKONPUVOVVNSU-UHFFFAOYSA-N
naphthalen-1-yl acetate
13247
Crystalline Powder or Crystals
Authentic
Pink or White to Yellow
100g
830-81-9
100.0
C12H10O2
MFCD00003922
1-acetoxynaphthalene, 1-naphthalenol, acetate, 1-naphthol, acetate, 1-naphthyl acetate, a-naphthyl acetate, acetic acid 1-naphthyl ester, alpha-naphthol acetate, alpha-naphthyl acetate, naphthalene, 1-acetoxy, naphthyl acetate
CC(=O)OC1=CC=CC2=CC=CC=C21
186.21
186.21
99%

Chemical Identifiers

830-81-9
186.21
VGKONPUVOVVNSU-UHFFFAOYSA-N
13247
CC(=O)OC1=CC=CC2=CC=CC=C21
C12H10O2
MFCD00003922
1-acetoxynaphthalene, 1-naphthalenol, acetate, 1-naphthol, acetate, 1-naphthyl acetate, a-naphthyl acetate, acetic acid 1-naphthyl ester, alpha-naphthol acetate, alpha-naphthyl acetate, naphthalene, 1-acetoxy, naphthyl acetate
naphthalen-1-yl acetate
Safety and Handling

Safety and Handling

EINECSNumber : 212-599-8

RTECSNumber : QL2975500

TSCA : TSCA

SDS Documents