missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
1-Naphthyl acetate, 99%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific 227250250
Specifications
1-Naphthyl acetate | |
Glass bottle | |
43.0°C to 46.0°C | |
99+% | |
99.0 | |
98.5% min. (HPLC) | |
CH3CO2C10H7 | |
06, 608 | |
VGKONPUVOVVNSU-UHFFFAOYSA-N | |
naphthalen-1-yl acetate | |
13247 | |
Crystalline Powder or Crystals |
Authentic | |
Pink or White to Yellow | |
25g | |
830-81-9 | |
100.0 | |
C12H10O2 | |
MFCD00003922 | |
1-acetoxynaphthalene, 1-naphthalenol, acetate, 1-naphthol, acetate, 1-naphthyl acetate, a-naphthyl acetate, acetic acid 1-naphthyl ester, alpha-naphthol acetate, alpha-naphthyl acetate, naphthalene, 1-acetoxy, naphthyl acetate | |
CC(=O)OC1=CC=CC2=CC=CC=C21 | |
186.21 | |
186.21 | |
99% |
Chemical Identifiers
830-81-9 | |
186.21 | |
VGKONPUVOVVNSU-UHFFFAOYSA-N | |
13247 | |
CC(=O)OC1=CC=CC2=CC=CC=C21 |
C12H10O2 | |
MFCD00003922 | |
1-acetoxynaphthalene, 1-naphthalenol, acetate, 1-naphthol, acetate, 1-naphthyl acetate, a-naphthyl acetate, acetic acid 1-naphthyl ester, alpha-naphthol acetate, alpha-naphthyl acetate, naphthalene, 1-acetoxy, naphthyl acetate | |
naphthalen-1-yl acetate |
Safety and Handling
EINECSNumber : 212-599-8
RTECSNumber : QL2975500
TSCA : TSCA