missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Naphthoxyacetic acid, 98+%

Manufacturer:  Alfa Aesar™ L0103606

 View more versions of this product

Catalog No. AAL0103606



Specifications

Specifications

1-Naphthoxyacetic acid
5g
C12H10O3
1912165
GHRYSOFWKRRLMI-UHFFFAOYSA-N
2-naphthalen-1-yloxyacetic acid
76313
202.21
193°C to 197°C
2976-75-2
MFCD00003927
1-naphthoxy acetic acid, 1-naphthoxyacetic acid, 1-naphthyloxy acetic acid, 1-naphthyloxyacetic acid, 2-1-naphthyloxy acetic acid, 2-naphthalen-1-yloxy acetic acid, acetic acid, 1-naphthalenyloxy, naphthalen-1-yloxy acetic acid, naphthyloxyacetic acid, unii-9qi2464poa
C1=CC=C2C(=C1)C=CC=C2OCC(=O)O
202.209
CHEBI:44588
≥98%

Chemical Identifiers

2976-75-2
202.209
GHRYSOFWKRRLMI-UHFFFAOYSA-N
76313
2-naphthalen-1-yloxyacetic acid
C12H10O3
MFCD00003927
1-naphthoxy acetic acid, 1-naphthoxyacetic acid, 1-naphthyloxy acetic acid, 1-naphthyloxyacetic acid, 2-1-naphthyloxy acetic acid, 2-naphthalen-1-yloxy acetic acid, acetic acid, 1-naphthalenyloxy, naphthalen-1-yloxy acetic acid, naphthyloxyacetic acid, unii-9qi2464poa
CHEBI:44588
C1=CC=C2C(=C1)C=CC=C2OCC(=O)O
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P264b-P270-P301+P312-P330-P501c

H302

EINECSNumber : 221-023-4

TSCA : No

Recommended Storage : Ambient temperatures

Documents