Please login to your online account to display your discounted pricing

1-Naphthoxyacetic acid, 97%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 1-Naphthoxyacetic acid
Name Note 97%
CAS 2976-75-2
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C12H10O3
Linear Formula C10H7OCH2COOH
Formula Weight 202.21
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC338920250

  SDS
thermo fisher scientific
338920250
Glass bottle 25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥96.0%
Physical Form Crystalline Powder
Formula Weight 202.21
Quantity 25g
Color Off-white to brown
TSCA TSCA
EINECS Number 221-023-4
Melting Point 193° to 198°C
CAS Min % 96.0
CAS Max % 100.0
Health Hazard 3 GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Packaging Glass bottle
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Infrared Spectrum Authentic
Molecular Formula C12H10O3
Assay Percent Range 97%
Linear Formula C10H7OCH2COOH
CAS 2976-75-2
Chemical Name or Material 1-Naphthoxyacetic acid
Name Note 97%
MDL Number MFCD00003927
Safety and Handling

GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.