missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Naphthonitrile, TCI America™

$96.25 - $478.20

Chemical Identifiers

CAS 86-53-3
Molecular Formula C11H7N
Molecular Weight (g/mol) 153.184
MDL Number MFCD00003866
InChI Key YJMNOKOLADGBKA-UHFFFAOYSA-N
Synonym 1-naphthonitrile, 1-cyanonaphthalene, 1-naphthalenecarbonitrile, naphthalenecarbonitrile, 1-naphthylnitrile, alpha-naphthonitrile, 1-naphthalenenitrile, alpha-naphthylnitrile, alpha-cyanonaphthalene, unii-2as1lz61hd
PubChem CID 6846
IUPAC Name naphthalene-1-carbonitrile
SMILES C1=CC=C2C(=C1)C=CC=C2C#N
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
N003825G View Documents TCI America
N003825G
25g Each for $96.25
 
N0038250G View Documents TCI America
N0038250G
250g Each for $478.20
 
Specifications

Chemical Identifiers

86-53-3
153.184
YJMNOKOLADGBKA-UHFFFAOYSA-N
6846
C1=CC=C2C(=C1)C=CC=C2C#N
C11H7N
MFCD00003866
1-naphthonitrile, 1-cyanonaphthalene, 1-naphthalenecarbonitrile, naphthalenecarbonitrile, 1-naphthylnitrile, alpha-naphthonitrile, 1-naphthalenenitrile, alpha-naphthylnitrile, alpha-cyanonaphthalene, unii-2as1lz61hd
naphthalene-1-carbonitrile

Specifications

299°C
25g
C11H7N
3439
YJMNOKOLADGBKA-UHFFFAOYSA-N
naphthalene-1-carbonitrile
6846
Crystal-Lump at 20°C
1-Naphthonitrile
White-Yellow
86-53-3
MFCD00003866
1-naphthonitrile, 1-cyanonaphthalene, 1-naphthalenecarbonitrile, naphthalenecarbonitrile, 1-naphthylnitrile, alpha-naphthonitrile, 1-naphthalenenitrile, alpha-naphthylnitrile, alpha-cyanonaphthalene, unii-2as1lz61hd
C1=CC=C2C(=C1)C=CC=C2C#N
153.184
153.18
≥95.0% (GC)
Safety and Handling

Safety and Handling

RTECSNumber : QL5976300

TSCA : Yes