missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Naphthoic Acid, TCI America™

$41.25 - $345.28

Chemical Identifiers

CAS 86-55-5
Molecular Formula C11H8O2
Molecular Weight (g/mol) 172.183
MDL Number MFCD00004007
InChI Key LNETULKMXZVUST-UHFFFAOYSA-N
Synonym 1-naphthoic acid, 1-naphthalenecarboxylic acid, 1-carboxynaphthalene, naphthoic acid, alpha-naphthoic acid, naphthalenecarboxylic acid, carboxynaphthalene, alpha-naphthylcarboxylic acid, naphthalene-alpha-carboxylic acid, unii-2niv4o66bh
PubChem CID 6847
ChEBI CHEBI:36466
IUPAC Name naphthalene-1-carboxylic acid
SMILES C1=CC=C2C(=C1)C=CC=C2C(=O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
N002425G View Documents TCI America
N002425G
25g Each for $41.25
 
N0024500G View Documents TCI America
N0024500G
500g Each for $345.28
 
Specifications

Chemical Identifiers

86-55-5
172.183
LNETULKMXZVUST-UHFFFAOYSA-N
6847
naphthalene-1-carboxylic acid
C11H8O2
MFCD00004007
1-naphthoic acid, 1-naphthalenecarboxylic acid, 1-carboxynaphthalene, naphthoic acid, alpha-naphthoic acid, naphthalenecarboxylic acid, carboxynaphthalene, alpha-naphthylcarboxylic acid, naphthalene-alpha-carboxylic acid, unii-2niv4o66bh
CHEBI:36466
C1=CC=C2C(=C1)C=CC=C2C(=O)O

Specifications

Undesignated
25g
C11H8O2
1-naphthoic acid, 1-naphthalenecarboxylic acid, 1-carboxynaphthalene, naphthoic acid, alpha-naphthoic acid, naphthalenecarboxylic acid, carboxynaphthalene, alpha-naphthylcarboxylic acid, naphthalene-alpha-carboxylic acid, unii-2niv4o66bh
C1=CC=C2C(=C1)C=CC=C2C(=O)O
172.183
CHEBI:36466
Crystal-Powder at 20°C
1-Naphthoic Acid
164°C
86-55-5
MFCD00004007
LNETULKMXZVUST-UHFFFAOYSA-N
naphthalene-1-carboxylic acid
6847
172.18
≥98.0% (T)
Safety and Handling

Safety and Handling

EINECSNumber : (4)-0397

RTECSNumber : QL0960000

TSCA : Yes