Learn More
1-Naphthoic acid, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1027314
Description
1-Naphthoic acid is used as an intermediate for the synthesis of pharmaceuticals, photochemicals, plant growth hormones, dyes and other organic compounds.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications1-Naphthoic acid is used as an intermediate for the synthesis of pharmaceuticals, photochemicals, plant growth hormones, dyes and other organic compounds.
Solubility
Freely soluble in hot alcohol and ether. Slightly soluble in hot water.
Notes
Store away from oxidizing agents. Store in a cool, dry condition in well sealed containers.
Specifications
1-Naphthoic acid | |
160°C to 162°C | |
>300°C | |
C11H8O2 | |
25 g | |
14,6381 | |
Freely soluble in hot alcohol and ether. Slightly soluble in hot water. | |
C1=CC=C2C(=C1)C=CC=C2C(=O)O | |
172.183 | |
CHEBI:36466 | |
98% |
86-55-5 | |
1.398 | |
195°C (383°F) | |
MFCD00004007 | |
1908896 | |
1-naphthoic acid, 1-naphthalenecarboxylic acid, 1-carboxynaphthalene, naphthoic acid, alpha-naphthoic acid, naphthalenecarboxylic acid, carboxynaphthalene, alpha-naphthylcarboxylic acid, naphthalene-alpha-carboxylic acid, unii-2niv4o66bh | |
LNETULKMXZVUST-UHFFFAOYSA-N | |
naphthalene-1-carboxylic acid | |
6847 | |
172.18 |
Chemical Identifiers
86-55-5 | |
172.183 | |
LNETULKMXZVUST-UHFFFAOYSA-N | |
6847 | |
naphthalene-1-carboxylic acid |
C11H8O2 | |
MFCD00004007 | |
1-naphthoic acid, 1-naphthalenecarboxylic acid, 1-carboxynaphthalene, naphthoic acid, alpha-naphthoic acid, naphthalenecarboxylic acid, carboxynaphthalene, alpha-naphthylcarboxylic acid, naphthalene-alpha-carboxylic acid, unii-2niv4o66bh | |
CHEBI:36466 | |
C1=CC=C2C(=C1)C=CC=C2C(=O)O |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 201-681-9
RTECSNumber : QL0960000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only