missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Naphthaleneboronic acid neopentyl glycol ester, 98%

$53.85 - $198.96

Chemical Identifiers

CAS 22871-77-8
Molecular Formula C15H17BO2
Formula Weight 240.11
MDL Number MFCD01318167
Beilstein 3053756
Chemical Name or Material 1-Naphthaleneboronic acid neopentyl glycol ester
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL1761203 SDS Alfa Aesar™
L1761203
1g Each for $53.85
AAL1761206 SDS Alfa Aesar™
L1761206
5g Each for $198.96
Specifications Description & Specifications

Chemical Identifiers

22871-77-8
240.11
3053756
C15H17BO2
MFCD01318167
1-Naphthaleneboronic acid neopentyl glycol ester

Specifications

67° to 69°C
22871-77-8
240.11
3053756
98%
1g
C15H17BO2
MFCD01318167
1-Naphthaleneboronic acid neopentyl glycol ester